missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Carboxypeptidase B Protein
A cDNA sequence encoding the Carboxypeptidase B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10544-50mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Carboxypeptidase B Protein | |
Research Use Only | |
170 units/mg protein. | |
Rat | |
Untagged | |
The protein was lyophilized with 100mM NaCl and 20mM Tris pH 7.5. |
50 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLLLLALVSV ALAHASEEHF DGNRVYRVSV HGEDHVNLIQ ELANTKEIDF WKPDSATQVK PLTTVDFHVK AEDVADVENF LEENEVHYEV LISNVRNALE SQFDSHTRAS GHSYTKYNKW ETIEAWIQQV ATDNPDLVTQ SVIGTTFEGR NMYVLKIGKT RPNKPAIFID CGFHAREWIS PAFCQWFVRE AVRTYNQEIH MKQLLDELDF YVLPVVNIDG YVYTWTKDRM WRKTRSTMAG SSCLGVRPNR NFNAGWCEVG ASRSPCSETY CGPAPESEKE TKALADFIRN NLSTIKAYLT IHSYSQMMLY PYSYDYKLPE NYEELNALVK GAAKELATLH GTKYTYGPGA TTIYPAAGGS DDWSYDQGIK YSFTFELRDT GFFGFLLPES QIRQTCEETM LAVKYIANYV REHLY | |
Greater than 90% as determined by SDS-PAGE. |