missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit TNC Protein
A cDNA sequence encoding the TNC was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13765-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
100009341 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Rabbit | |
MTDQQAEARS YLSEEMIAEF KAAFDMFDAD GGGDISVKEL GTVMRMLGQT PTKEELDAII EEVDEDGSGT IDFEEFLVMM VRQMKEDAKG KSEEELAECF RIFDRNADGY IDAEELAEIF RASGEHVTDE EIESLMKDGD KNNDGRIDFD EFLKMMEGVQ | |
Greater than 95.0% as determined by SDS-PAGE. |
TNC Protein | |
Research Use Only | |
TNNC2 | |
Rabbit | |
Untagged | |
The Troponin C Rabbit protein solution contains 150mM sodium chloride, 10mM sodium phosphate and 0.05% sodium azide, pH 7.2. |