missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 2750.00 CHF
Spezifikation
Name | IGF1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | IGF-I is biologically active when compared to human IGF-1. The ED50, calculated by the dose -dependent proliferation of human MCF/7 cells is 5 to 25ng/ml in the cell culture mixture dependent on culture conditions. Its activity consists of 30-40 % compared to human IGF-1. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15944628
|
enQuireBio™
QP10679-10UG |
10 μg |
131.00 CHF
10 Mikrogramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964628
|
enQuireBio™
QP10679-50UG |
50 μg |
203.00 CHF
50 Mikrogramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15954628
|
enQuireBio™
QP10679-1MG |
1 mg |
2750.00 CHF
1 Milligramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
IGF1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MTAGPETLCG AELVDALQFV CGDRGFYFNK PTGYGSSSRR APQTGIVDEC CFRSCDLRRL EMYCAPLKP | |
Greater than 98.0% as determined by:(a) Gel-filtration chromatography under non denaturing conditions. (b) Analysis by SDS-PAGE. |
Research Use Only | |
IGF-I is biologically active when compared to human IGF-1. The ED50, calculated by the dose -dependent proliferation of human MCF/7 cells is 5 to 25ng/ml in the cell culture mixture dependent on culture conditions. Its activity consists of 30-40 % compared to human IGF-1. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (0.5 mg/ml) solution with 0.02% NaHCO3. |