missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine OXM Protein
A cDNA sequence encoding the OXM was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 2025.00 CHF
Spezifikation
Name | OXM Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Porcine |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15902029
|
enQuireBio™
QP12942-10UG |
10 μg |
131.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15922029
|
enQuireBio™
QP12942-50UG |
50 μg |
203.00 CHF
50 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15912029
|
enQuireBio™
QP12942-1MG |
1 mg |
2025.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
OXM Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-Lys-Arg-Asn-Lys-Asn-Asn-Ile-Ala | |
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |