missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other SSBP1 Protein

A cDNA sequence encoding the SSBP1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13592-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15964079

Menge Preis
1 192.32 CHF / 20 Mikrogramm
6 177.22 CHF / 20 Mikrogramm
Voraussichtliches Versanddatum: 10-11-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

SSBP1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
SSBP1 is a 0.2microm filtered concentrated solution which contains 20mM Tris, pH 7.4, 200mM NaCl, 1mM EDTA, 0.5mM DTT, and 50% Glycerol.
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MEEKVGNLKP NMESVNVTVR VLEASEARQI QTKNGVRTIS EAIVGDETGR VKLTLWGKHA GSIKEGQVVK IENAWTTAFK GQVQLNAGSK TKIAEASEDG FPESSQIPEN TPTAPQQMRG GGRGFRGGGR RYGRRGGRRQ ENEEGEEE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate