missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SPA Protein
A cDNA sequence encoding the SPA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13566-1gr
Additional Details : Gewicht : 0.01000kg
Spezifikation
SPA Protein | |
Research Use Only | |
spa | |
Other | |
His | |
SPA protein was lyophilized with no additives. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MNAAQHDEAQ QNAFYQVLNM PNLNADQRNG FIQSLKDDPS QSANVLGEAQ KLNDSQAPKA DAQQNNFNKD QQSAFYEILN MPNLNEAQRN GFIQSLKDDP SQSTNVLGEA KKLNESQAPK ADNNFNKEQQ NAFYEILNMP NLNEEQRNGF IQSLKDDPSQ SANLLSEAKK LNESQAPKAD NKFNKEQQNA FYEILHLPNL NEEQRNGFIQ SLKDDPSQSA NLLAEAKKLN DAQAPKADNK FNKEQQNAFY EILHLPNLTE EQRNGFIQSL KDDPSVSKEI LAEAKKLNDA QAPKEEDSLE HHHHHH | |
Greater than 97.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |