missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SKP Protein
A cDNA sequence encoding the SKP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13516-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SKP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADKIAIVNM GSLFQQVAQK TGVSNTLENE FKGRASELQR METDLQAKMK KLQSMKAGSD RTKLEKDVMA QRQTFAQKAQ AFEQDRARRS NEERGKLVTR IQTAVKSVAN SQDIDLVVDA NAVAYNSSDV KDITADVLKQ VK | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
SKP E. Coli solution containing 20mM Tris-HCl pH 8 and 20% glycerol. |