missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Shiga Like Toxin 1 Protein
A cDNA sequence encoding the Shiga Like Toxin 1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13495-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Shiga Like Toxin 1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline and 50mM arginine, pH 7.4. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |