missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Shiga Like Toxin 1 Protein
A cDNA sequence encoding the Shiga Like Toxin 1 was constructed and used to recombinantly synthesize the protein.
227.00 CHF - 1735.00 CHF
Spezifikation
Name | Shiga Like Toxin 1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Other |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983749
|
enQuireBio™
QP13495-100UG |
100 μg |
227.00 CHF
100 Mikrogramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15903759
|
enQuireBio™
QP13495-500UG |
500 μg |
870.00 CHF
500 Mikrogramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15993749
|
enQuireBio™
QP13495-1MG |
1 mg |
1735.00 CHF
1 Milligramm |
Verfügbar ab: 20-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Shiga Like Toxin 1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline and 50mM arginine, pH 7.4. |