missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Protein-L Protein

A cDNA sequence encoding the Protein-L was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13168-100mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15962709

Menge Preis
1 852.02 CHF / 100 Milligramm
5 783.52 CHF / 100 Milligramm
Voraussichtliches Versanddatum: 28-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

Protein-L Protein
100 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein-L was lyophilized with no additives.
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
KEETPETPET DSEEEVTIKA NLIFANGSTQ TAEFKGTFEK ATSEAYAYAD TLKKDNGEYT VDVADKGYTL NIKFAGKEKT PEEPKEEVTI KANLIYADGK TQTAEFKGTF EEATAEAYRY ADALKKDNGE YTVDVADKGY TLNIKFAGKE KTPEEPKEEV TIKANLIYAD GKTQTAEFKG TFEEATAEAY RYADLLAKEN GKYTVDVADK GYTLNIKFAG KEKTPEEPKE EVTIKANLIY ADGKTQTAEF KGTFAEATAE AYRYADLLAK ENGKYTADLE DGGYTINIRF AGKKVDEKPEEKEQVTIKEN IYFEDGTVQT ATFKGTFAEA TAEAYRYADL LSKEHGKYTA DLEDGGYTIN IRFAG
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate