missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other protein 1
A cDNA sequence encoding the Protein G was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13162-100mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
protein 1 | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
LPKTDTYKLILNGKTLKGETTTEAVDAATAEKVFKQYANDNGVDGEWTYDDAT KTFTVTEKPEVIDASELTPAVTTYKLVINGKTLKGETTTEAVDAATAEKVFK QYANDNGVDGEWTYDDATKTFTVTEKPEVIDASELTPAVTTYKLVINGKTL KGETTTKAVDAETAEKAFKQYANDNGVDGVWTYDDATKTFTVTE | |
>96% as determined by SDS-PAGE and RP-HPLC. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Lyophilized white powder containing no additives. |