missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PhI p 5 Protein
A cDNA sequence encoding the PhI p 5 was constructed and used to recombinantly synthesize the protein.
870.00 CHF - 5790.00 CHF
Spezifikation
Name | PhI p 5 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Other |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15962329
|
enQuireBio™
QP13042-50UG |
50 μg |
870.00 CHF
50 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15942329
|
enQuireBio™
QP13042-100UG |
100 μg |
1300.00 CHF
100 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15952329
|
enQuireBio™
QP13042-1MG |
1 mg |
5790.00 CHF
1 Milligramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PhI p 5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
PhI p 5 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
SVKRSNGSAE VHRGAVPRRG PRGGPGRSYA ADAGYAPATP AAAGAEAGKA TTEEQKLIED INVGFKAAVA AAASVPAADK FKTFEAAFTS SSKAATAKAP GLVPKLDAAY SVAYKAAVGA TPEAKFDSFV ASLTEALRVI AGALEVHAVK PVTEEPGMAK IPAGELQIID KIDAAFKVAA TAAATAPADD KFTVFEAAFN KAIKESTGGA YDTYKCIPSL EAAVKQAYAA TVAAAPQVKY AVFEAALTKA ITAMSEVQKV SQPATGAATV AAGAATTAAG AASGAATVAA GGYKV | |
Greater than 80.0% as determined by SDS-PAGE. |