missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PhI p 1 Protein
A cDNA sequence encoding the PhI p 1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13040-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PhI p 1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MASSSSVLLV VVLFAVFLGS AYGIPKVPPG PNITATYGDK WLDAKSTWYG KPTGAGPKDN GGACGYKDVD KPPFSGMTGC GNTPIFKSGR GCGSCFEIKC TKPEACSGEP VVVHITDDNE EPIAPYHFDL SGHAFGAMAK KGDEQKLRSA GELELQFRRV KCKYPEGTKV TFHVEKGSNP NYLALLVKYV NGDGDVVAVD IKEKGKDKWI ELKESWGAIW RIDTPDKLTG PFTVRYTTEG GTKTEAEDVI PEGWKADTSY ESK | |
Greater than 80.0% as determined by SDS-PAGE. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
PhI p 1 protein solution is supplied in 20mM HEPES pH 8.0, 200mM NaCl and 20% glycerol. |