missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other melA Protein
A cDNA sequence encoding the melA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12676-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
948636 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMSAPKI TFIGAGSTIF VKNILGDVFH REALKTAHIA LMDIDPTRLE ESHIVVRKLM DSAGASGKIT CHTQQKEALE DADFVVVAFQ IGGYEPCTVT DFEVCKRHGL EQTIADTLGP GGIMRALRTI PHLWQICEDM TEVCPDATML NYVNPMAMNT WAMYARYPHI KQVGLCHSVQ GTAEELARDL NIDPATLRYR CAGINHMAFY LELERKTADG SYVNLYPELL AAYEAGQAPK PNIHGNTRCQ NIVRYEMFKK LGYFVTESSE HFAEYTPWFI KPGREDLIER YKVPLDEYPK RCVEQLANWH KELEEYKKAS RIDIKPSREY ASTIMNAIWT GEPSVIYGNV RNDGLIDNLP QGCCVEVACL VDANGIQPTK VGTLPSHLAA LMQTNINVQT LLTEAILTEN RDRVYHAAMM DPHTAAVLGI DEIYALVDDL IAAHGDWLPG WLHR | |
Greater than 90% as determined by SDS-PAGE. |
melA Protein | |
Research Use Only | |
melA | |
Other | |
His | |
The melA solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol. |