missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lanreotide Protein
A cDNA sequence encoding the Lanreotide was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12536-10mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Lanreotide Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with 5 mg manntitol and 0.04 mg tween-80. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |