missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HSP104 Saccharomyces Protein

A cDNA sequence encoding the HSP104 Saccharomyces was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12325-50ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910069

  • 203.00 CHF / 50 Mikrogramm
Verfügbar ab: 29-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

HSP104 Saccharomyces Protein
Research Use Only
Recombinant Protein
S. cerevisiae
MNDQTQFTER ALTILTLAQK LASDHQHPQL QPIHILAAFI ETPEDGSVPY LQNLIEKGRY DYDLFKKVVN RNLVRIPQQQ PAPAEITPSY ALGKVLQDAA KIQKQQKDSF IAQDHILFAL FNDSSIQQIF KEAQVDIEAI KQQALELRGN TRIDSRGADT NTPLEYLSKY AIDMTEQARQ GKLDPVIGRE EEIRSTIRVL ARRIKSNPCL IGEPGIGKTA IIEGVAQRII DDDVPTILQG AKLFSLDLAA LTAGAKYKGD FEERFKGVLK EIEESKTLIV LFIDEIHMLM GNGKDDAANI LKPALSRGQL KVIGATTNNE YRSIVEKDGA FERRFQKIEV AEPSVRQTVA ILRGLQPKYE IHHGVRILDS ALVTAAQLAK RYLPYRRLPD SALDLVDISC AGVAVARDSK PEELDSKERQ LQLIQVEIKA LERDEDADST TKDRLKLARQ KEASLQEELE PLRQRYNEEK HGHEELTQAK KKLDELENKA LDAERRYDTA TAADLRYFAI PDIKKQIEKL EDQVAEEERR AGANSMIQNV VDSDTISETA ARLTGIPVKK LSESENEKLI HMERDLSSEV VGQMDAIKAV SNAVRLSRSG LANPRQPASF LFLGLSGSGK TELAKKVAGF LFNDEDMMIR VDCSELSEKY AVSKLLGTTA GYVGYDEGGF LTNQLQYKPY SVLLFDEVEK AHPDVLTVML QMLDDGRITS GQGKTIDCSN CIVIMTSNLG AEFINSQQGS KIQESTKNLV MGAVRQHFRP EFLNRISSIV IFNKLSRKAI HKIVDIRLKE IEERFEQNDK HYKLNLTQEA KDFLAKYGYS DDMGARPLNR LIQNEILNKL ALRILKNEIK DKETVNVVLK KGKSRDENVP EEAEECLEVL PNHEATIGAD TLGDDDNEDS MEIDDDLD
Greater than 90.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The HSP-104 protein solution contains 20mM Tris-HCl, pH 7.4,100mM NaCl, 2mM EDTA and 5% Glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote