Learn More
enQuireBio™ Recombinant other HPSE Active Protein
Recombinant Protein
Marke: enQuireBio™ QP10668-4ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
HPSE Active Protein | |
Research Use Only | |
The specific activity of Heparanase Active Enzyme in-house standard is about 0.7 Units (1 unit = 1 umole of reducing ends of heparan sulfate substrate formed per minute per mg Heparanase Active Enzyme at 37C). The enzymatic activity of each Heparanase Active Enzyme batch is comparable to the standard as determined by activity assay in which immobilized heparan, released due to heparanase activity, is quantified colorimetrically. Recommended reaction buffer: 20 mM Citrate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2. | |
Other | |
rate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2. |
4 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Untagged | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |