missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GST Protein

A cDNA sequence encoding the GST was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10656-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15964538

Menge Preis
1 128.22 CHF / 10 Mikrogramm
10 117.77 CHF / 10 Mikrogramm
Voraussichtliches Versanddatum: 28-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

GST Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MSPILGYWKI KGLVQPTRLL LEYLEEKYEE HLYERDEGDK WRNKKFELGL EFPNLPYYID GDVKLTQSMA IIRYIADKHN MLGGCPKERA EISMLEGAVL DIRYGVSRIA YSKDFETLKV DFLSKLPEML KMFEDRLCHK TYLNGDHVTH PDFMLYDALD VVLYMDPMCL DAFPKLVCFK KRIEAIPQID KYLKSSKYIA WPLQGWQATF GGGDHPPKSD LVPR
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Specific activity is >20 units/mg. A unit is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C.
Other
Untagged
GST supplied in Phosphate Buffered Saline pH 7.4.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate