missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other CK2a Zea Mays Protein
A cDNA sequence encoding the CK2a Zea Mays was constructed and used to recombinantly synthesize the protein.
580.00 CHF - 2170.00 CHF
Spezifikation
Name | CK2a Zea Mays Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Other |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15957248
|
enQuireBio™
QP11433-3UG |
3 μg |
580.00 CHF
3 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15937248
|
enQuireBio™
QP11433-10UG |
10 μg |
1160.00 CHF
10 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15947248
|
enQuireBio™
QP11433-20UG |
20 μg |
2170.00 CHF
20 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
CK2a Zea Mays Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
CK2a is supplied in 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSKARVYADV NVLRPKEYWD YEALTVQWGE QDDYEVVRKV GRGKYSEVFE GINVNNNEKC IIKILKPVKK KKIKREIKIL QNLCGGPNIV KLLDIVRDQH SKTPSLIFEY VNNTDFKVLY PTLTDYDIRY YIYELLKALD YCHSQGIMHR DVKPHNVMID HELRKLRLID WGLAEFYHPG KEYNVRVASR YFKGPELLVD LQDYDYSLDM WSLGCMFAGM IFRKEPFFYG HDNHDQLVKI AKVLGTDGLN VYLNKYRIEL DPQLEALVGR HSRKPWLKFM NADNQHLVSP EAIDFLDKLL RYDHQERLTA LEAMTHPYFQ QVRAAENSRT RA | |
Greater than 99% as determined by SDS-PAGE. |