missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mumps Virus Mumps Protein

A cDNA sequence encoding the Mumps was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12753-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911439

  • 1660.00 CHF / 1 Milligramm


Verfügbar ab
11-09-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

Mumps Virus Mumps Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mumps Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Phosphate buffer, pH 7.4.
Videos
Dokumentation