missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mumps Virus Mumps Protein
A cDNA sequence encoding the Mumps was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12753-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Mumps Virus Mumps Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffer, pH 7.4. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mumps Virus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |