missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species PKACa2-RIIa2 Protein
A cDNA sequence encoding the PKACa2-RIIa2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5355.00 CHF
Spezifikation
Name | Multi Species PKACa2-RIIa2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15975278
|
enQuireBio™
QP10824-1UG |
1 μg |
131.00 CHF
1 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15985278
|
enQuireBio™
QP10824-3UG |
3 μg |
203.00 CHF
3 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965278
|
enQuireBio™
QP10824-100UG |
100 μg |
5355.00 CHF
100 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Multi Species PKACa2-RIIa2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
PKA holoenzyme type-II alpha is supplied in 50% glycerol. |
Research Use Only | |
Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits. | |
Multi Species | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |