missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species PKA-RII alpha Protein
A cDNA sequence encoding the PKA-RII alpha was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13063-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Multi Species PKA-RII alpha Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSHIQIPPGL TELLQGYTVE VLRQQPPDLV EFAVEYFTRL REARAPASVL PAATPRQSLG HPPPEPGPDR VADAKGDSES EEDEDLEVPV PSRFNRRVSV CAETYNPDEE EEDTDPRVIH PKTDEQRCRL QEACKDILLF KNLDQEQLSQ VLDAMFERIV KADEHVIDQG DDGDNFYVIE RGTYDILVTK DNQTRSVGQY DNRGSFGELA LMYNTPRAAT IVATSEGSLW GLDRVTFRRI IVKNNAKKRK MFESFIESVP LLKSLEVSER MKIVDVIGEK IYKDGERIIT QGEKADSFYI IESGEVSILI RSRTKSNKDG GNQEVEIARC HKGQYFGELA LVTNKPRAAS AYAVGDVKCL VMDVQAFERL LGPCMDIMKR NISHYEEQLV KMFGSSVDLG NLGQ | |
Greater than 95% as determined by SDS-PAGE. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Multi Species | |
Untagged | |
PKA regulatory subunit-II alpha is supplied in 20mM MES, 150mM NaCl, 2mM EDTA, 2mM EGTA, 1mM BME and 50% glycerol. |