missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNF-alpha Protein
A DNA sequence encoding the Mouse TNF-alpha, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10470-ec-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001265530.1 | |
TNF-alpha Protein | |
Research Use Only | |
TNF | |
The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg in the presence of actinomycin D. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.2, lyophilized |
21926 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL | |
Greater than 98% as determined by SDS-PAGE and HPLC. |