missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TGF-beta 1 Protein
Click to view available options
Zusammensetzung:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Packungsgröße:
5 Mikrogramm
Beschreibung
The Recombinant Mouse TGF-beta 1 Protein is derived from CHO. The Recombinant Mouse TGF-beta 1 Protein has been validated for the following applications: Bioactivity.

Spezifikation
Spezifikation
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
Gen-ID (Entrez) | 21803 |
Molekulargewicht | 12.8 kDa (monomer) |
Name | TGF-beta 1 |
Menge | 5 μg |
Quelle | Chinese Hamster Ovary cell line,CHO-derived mouse TGF-beta 1 protein Ala279-Ser390 |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Strukturform | Disulfide-linked homodimer |
Gen-Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
Konjugat | Unconjugated |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse TGF-beta 1 Protein >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur