missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SNCG Protein
A cDNA sequence encoding the SNCG was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13537-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SNCG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDVFKKG FSIAKEGVVG AVEKTKQGVT EAAEKTKEGV MYVGTKTKEN VVQSVTSVAE KTKEQANAVS AVVSSVNTV ANKTVEEAEN IVVTTGVVRK EDLEPPAQDQ EAKEQEENEE AKSGED | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
SNCG protein solution (1 mg/ml) containing 20mM Tris-HCl(pH 8.0) and 0.1M NaCl. |