missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SF20 Protein

A cDNA sequence encoding the SF20 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13475-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983689

  • 188.00 CHF / 10 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

SF20 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
Untagged
Mouse SF20 was lyophilized from a 0.2microm filtered concentrated (1.0 mg/ml) solution in PBS, pH 7.4.
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL
Videos
Dokumentation