missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SCF / C-kit ligand Protein

A DNA sequence encoding the Mouse SCF / C-kit ligand, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10439-ec-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983858

  • 123.00 CHF / 2 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001334085.1
SCF / C-kit ligand Protein
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
17311
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
Kitlg
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation