missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A1 Protein
A cDNA sequence encoding the S100A1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13386-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
S100A1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSELESAME TLINVFHAHS GKEGDKYKLS KKELKDLLQT ELSGFLDVQK DADAVDKVMK ELDENGDGEV DFKEYVVLVA ALTVACNNFF WETS | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
S100A1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol and 0.1M NaCl. |