missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Prolactin / PRL Protein

A DNA sequence encoding the Mouse Prolactin / PRL, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10153-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982908

  • 1155.00 CHF / 250 Mikrogramm


Verfügbar ab
29-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

Prolactin / PRL Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
LPICSAGDCQ TSLRELFDRV VILSHYIHTL YTDMFIEFDK QYVQDREFMV KVINDCPTSS LATPEDKEQA LKVPPEVLLN LILSLVQSSS DPLFQLITGV GGIQEAPEYI LSRAKEIEEQ NKQLLEGVEK IISQAYPEAK GNGIYFVWSQ LPSLQGVDEE SKILSLRNTI RCLRRDSHKV DNFLKVLRCQ IAHQNNC
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
PRL
The ED50 as determined by a cell proliferation assay using rat Nb2-11 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation