missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PRDX2 Protein

A cDNA sequence encoding the PRDX2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10500-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15984058

  • 7190.00 CHF / 1 Milligramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

PRDX2 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMASGNA QIGKSAPDFT ATAVVDGAFK EIKLSDYRGK YVVLFFYPLD FTFVCPTEII AFSDHAEDFR KLGCEVLGVS VDSQFTHLAW INTPRKEGGL GPLNIPLLAD VTKSLSQNYG VLKNDEGIAY RGLFIIDAKG VLRQITVNDL PVGRSVDEAL RLVQAFQYTD EHGEVCPAGW KPGSDTIKPN VDDSKEYFSK HN
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 700 pmol/min/ug, Activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for minute.
Mouse
His
PRDX2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol 1mM DTT.
Videos
Dokumentation