missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse OGG1 Protein
A cDNA sequence encoding the OGG1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12922-1ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
OGG1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLFRSWL PSSMRHRTLS SSPALWASIP CPRSELRLDL VLASGQSFRW KEQSPAHWSG VLADQVWTLT QTEDQLYCTV YRGDDSQVSR PTLEELETLH KYFQLDVSLA QLYSHWASVD SHFQRVAQKF QGVRLLRQDP TECLFSFICS SNNNIARITG MVERLCQAFG PRLIQLDDVT YHGFPNLHAL AGPEAETHLR KLGLGYRARY VRASAKAILE EQGGPAWLQQ LRVAPYEEAH KALCTLPGVG AKVADCICLM ALDKPQAVPV DVHVWQIAHR DYGWHPKTSQ AKGPSPLANK ELGNFFRNLW GPYAGWAQAV LFSADLRQPS LSREPPAKRK KGSKRPEG | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
OGG1 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 30% glycerol. |