missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse NKp46 Protein

A cDNA sequence encoding the NKp46 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12851-1ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911749

  • 131.00 CHF / 1 Mikrogramm
Verfügbar ab: 28-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

NKp46 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADLQRINTEK ETLPKPIIWA KPSIMVTNGN SVNIWCQGAQ SASEYQLYFE GSFFALERPK PSRSMNKVRF FISQMTSHTA GIYTCFYQSG ELWSKSSNPL KLVVTGLYDT PNLWVYPRPE VTLGENVTFF CQLKTATSKF FLLKERGSNH IQNKYGNIQA EFPMGPVTRA HRGTYRCFGSYNDYAWSFPS EPVTLLITGG VENSSLAPTD PTSSLDYWEF DLSTNESGLQ KDSAFWDHTT QNLEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPIEKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H
Greater than 85.0% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
NKp46 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote