missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MIF Protein
Click to view available options
Zusammensetzung:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Packungsgröße:
25 Mikrogramm
Beschreibung
The Recombinant Mouse MIF Protein is derived from E. coli. The Recombinant Mouse MIF Protein has been validated for the following applications: Bioactivity.

Spezifikation
Spezifikation
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS. |
Gen-ID (Entrez) | 17319 |
Molekulargewicht | 12.4 kDa |
Name | MIF |
Menge | 25 μg |
Quelle | E. coli-derived mouse MIF protein Pro2-Ala115 |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Konjugat | Unconjugated |
Rekombinant | Recombinant |
Reinheits- oder Qualitätsgrad | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse MIF Protein >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur