missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse M-CSF Protein
Click to view available options
Menge:
10 μg
50 μg
500 μg
Zusammensetzung:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Packungsgröße:
10 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
The Recombinant Mouse M-CSF Protein is derived from E. coli. The Recombinant Mouse M-CSF Protein has been validated for the following applications: Bioactivity.

Spezifikation
Spezifikation
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS. |
Gen-ID (Entrez) | 12977 |
Molekulargewicht | 26 kDa (monomer) |
Name | M-CSF |
Menge | 50 μg |
Quelle | E. coli-derived mouse M-CSF protein Lys33-Glu262,with an N-terminal Met |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Strukturform | Disulfide-linked homodimer |
Konjugat | Unconjugated |
Rekombinant | Recombinant |
Mehr anzeigen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse M-CSF Protein >
Spot an opportunity for improvement?Share a Content Correction