missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interleukin-2 / IL-2 Protein
A DNA sequence encoding the Mouse Interleukin-2 / IL-2 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10451-ec-250ug
Dettagli aggiuntivi : Gewicht : 0.01000kg
Specifica
NP_032392.1 | |
Interleukin-2 / IL-2 Protein | |
Research Use Only | |
IL2 | |
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.2 ng/ml, corresponding to a specific activity of > 5.0 M 106 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16183 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APTSSSTSSS TAEAQQQQQQ QQQQQQHLEQ LLMDLQELLS RMENYRNLKL PRMLTFKFYL PKQATELKDL QCLEDELGPL RHVLDLTQSK SFQLEDAENF ISNIRVTVVK LKGSDNTFEC QFDDESATVV DFLRRWIAFC QSIISTSPQ | |
Greater than 95% as determined by SDS-PAGE and HPLC. |