missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein
Click to view available options
Menge:
50 μg
Packungsgröße:
50 Mikrogramm
Beschreibung
Includes
For Research Use Only
The Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein has been validated for the following applications: Bioactivity.
Bestellinformation
Ships at ambient temperature.

Spezifikation
Spezifikation
Zugriffsnummer | Q62470-1 (Integrin alpha 3) & P09055-1 (Integrin beta 1) |
Zur Verwendung mit (Anwendung) | Bioactivity |
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS |
Gen-ID (Entrez) | 3675 |
Molekulargewicht | 116 kDa (Integrin alpha 3), 86 kDa (Integrin beta 1) |
Menge | 50 μg |
Quelle | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 3 beta 1/VLA-3 protein, (N-terminus) Mouse Integrin alpha 3 (Phe33-Glu993) Accession &Num; Q62470-1 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055-1 His-Pro GGGSGGGS Basic Tail (C-terminus) |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Kennzeichnung | RUO |
Endotoxin-Konzentration | <0.10 EU per 1μg of the protein by the LAL method. |
Mehr anzeigen |
Produktvorschläge
Customers who viewed this item also viewed.
Viewing 1-4 of
For Research Use Only.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur