missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL7 / Interleukin 7 Protein

A DNA sequence encoding the Mouse IL7 / Interleukin 7 Protein, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10456-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983918

  • 4010.00 CHF / 250 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_032397.1
IL7 / Interleukin 7 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ECHIKDKEGK AYESVLMISI DELDKMTGTD SNCPNNEPNF FRKHVCDDTK EAAFLNRAAR KLKQFLKMNI SEEFNVHLLT VSQGTQTLVN CTSKEEKNVK EQKKNDACFL KRLLREIKTC WNKILKGSI
16196
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
IL7
The ED50 as determined by a cell proliferation assay using murine 2E8 cells is less than 0.2 ng/ml, corresponding to a specific activity of > 5.0 M 106 IU/mg.
Mouse
Untagged
0.2 um filtered PBS, pH 7.4, 2 % trehalose, lyophilized
Videos
Dokumentation