missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL6R Protein
A cDNA sequence encoding the IL6R was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12428-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
16195 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
QLLEPCGYIY PEFPVVQRGS NFTAICVLKE ACLQHYYVNA SYIVWKTNHA AVPREQVTVI NRTTSSVTFT DVVLPSVQLT CNILSFGQIE QNVYGVTMLS GFPPDKPTNL TCIVNEGKNM LCQWDPGRET YLETNYTLKS EWATEKFPDC QSKHGTSCMV SYMPTYYVNI EVWVEAENAL GKVSSESINF DPVDKVKPTP PYNLSVTNSE ELSSILKLSW VSSGLGGLLD LKSDIQYRTK DASTWIQVPL EDTMSPRTSF TVQDLKPFTE YVFRIRSIKD SGKGYWSDWS EEASGTTYED RPSRPPSFWY KTNPSHGQEY RSVRLIWKAL PLSEANGKIL DYEVILTQSK SVSQTYTVTG TELTVNLTND RYVASLAARN KVGKSAAAVL TIPSPHVTAA YSVVNLKAFP KDNLLWVEWT PPPKPVSKYI LEWCVLSENA PCVEDWQQED ATVNRTHLRG RLLESKCYQI TVTPVFATGP GGSESLKAYL KQAAPARGPT VRTKKVGKNE AVLAWDQIPV DDQNGFIRNY SISYRTSVGK EMVVHVDSSH TEYTLSSLSS DTLYMVRMAA YTDEGGKDGP EFTFTTPKFA QGEIELEHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL6R Protein | |
Research Use Only | |
Il6st | |
Mouse | |
His | |
IL6R protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |