missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL33 / Interleukin-33 / NF-HEV Protein
A DNA sequence encoding the Mouse IL33 / Interleukin-33 / NF-HEV Protein, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10464-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001158196.1 | |
IL33 / Interleukin-33 / NF-HEV Protein | |
Research Use Only | |
IL33 | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, and 1 mM EDTA, lyophilized |
77125 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SIQGTSLLTQ SPASLSTYND QSVSFVLENG CYVINVDDSG KDQEQDQVLL RYYESPCPAS QSGDGVDGKK LMVNMSPIKD TDIWLHANDK DYSVELQRGD VSPPEQAFFV LHKKSSDFVS FECKNLPGTY IGVKDNQLAL VEEKDESCNN IMFKLSKI | |
Greater than 98% as determined by SDS-PAGE and HPLC. |