missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL16 / Interleukin-16 Protein
A DNA sequence encoding the Mouse IL16 / Interleukin-16, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10461-ec-250ug
Detalles adicionales : Gewicht : 0.01000kg
Especificaciones
NP_034681.2 | |
IL16 / Interleukin-16 Protein | |
Research Use Only | |
IL16 | |
The biological activity determined by a chemotaxis bioassay using human peripheral T lymphocytes is in a concentration range of 1.0-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16170 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SAASASAASD ISVESKEATV CTVTLEKTSA GLGFSLEGGK GSLHGDKPLT INRIFKGTEQ GEMVQPGDEI LQLAGTAVQG LTRFEAWNVI KALPDGPVTI VIRRTSLQCK QTTASADS | |
Greater than 98% as determined by SDS-PAGE and HPLC. |