missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-5 R alpha/CD125 His-tag Protein
Carrier Free Recombinant Protein
Marke: R&D Systems™ 11367-5R-025
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its binding ability in a functional ELISA. When Recombinant Mouse IL-5 (Catalog # 405-ML ) is immobilized at 4.0μg/mL (100μL/well), Recombinant Mouse IL-5R alpha/CD125 His-tag (Catalog # 11367-5R) binds with an ED50 of 0.250-2.50μg/mL
Spezifikation
P21183.1 | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
M.W. (Observed): 48-54 kDa, under reducing conditions., M.W. (Theoretical): 38 kDa | |
Human embryonic kidney cell, HEK293-derived mouse IL-5 R alpha/CD125 protein Asp18-His339, with a C-terminal 6-His tag | |
<0.10 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100μg/mL in PBS. | |
IL-5R alpha/CD125 |
Bioactivity | |
3568 | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
Mouse | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse IL-5 R alpha/CD125 His-tag Protein > 25 μg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur