missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-28A/IFN-lambda 2 Protein
Click to view available options
Zusammensetzung:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Packungsgröße:
25 Mikrogramm
Beschreibung
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.0800-0.800 ng/mL.

Spezifikation
Spezifikation
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Molekulargewicht | 20.5 kDa |
Name | IL-28A/IFN-lambda 2 |
Menge | 25 μg |
Quelle | Mouse myeloma cell line,NS0-derived mouse IL-28A/IFN-lambda 2 protein Asp20-Val193,with a C-terminal 6-His tag |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Konjugat | Unconjugated |
Rekombinant | Recombinant |
Reinheits- oder Qualitätsgrad | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Mouse IL-28A/IFN-lambda 2 Protein >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur