missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-11 / interleukin 11 Protein
A DNA sequence encoding the Mouse IL-11 / interleukin 11, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10459-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_032376.1 | |
IL-11 / interleukin 11 Protein | |
Research Use Only | |
IL11 | |
The ED50 as determined by a cell proliferation assay using murine T11 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16156 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |