missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGFBP6 Protein
A cDNA sequence encoding the IGFBP6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12382-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
IGFBP6 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ALAGCPGCGA GMQTGCRGGC VEEEDAGSPA DGCTEAGGCL RREGQPCGVY SPKCAPGLQC QPRENEEAPL RALLIGQGRC QRARGPSEET TKESKPQGGA SRSRDTNHRD RQKNPRTSAA PIRPNPVQDS EMGPCRRHLD SVLQQLQTEV FRGGARGLYV PNCDLRGFYR KQQCRSSQGN RRGPCWCVDP MGQPLPVSPD GQGSTQCSAR SSGLEHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
IGFBP6 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 40% glycerol. |