missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFN b Protein
A cDNA sequence encoding the IFN b was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12374-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
15977 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MINYKQLQLQ ERTNIRKCQE LLEQLNGKIN LTYRADFKIP MEMTEKMQKS YTAFAIQEML QNVFLVFRNN FSSTGWNETI VVRLLDELHQ QTVFLKTVLE EKQEERLTWE MSSTALHLKS YYWRVQRYLK LMKYNSYAWM VVRAEIFRNF LIIRRLTRNF QN | |
Greater than 95% as determined by Analysis by SDS-PAGE. |
IFN b Protein | |
Research Use Only | |
Ifnb1 | |
Mouse | |
Untagged | |
The IFN-Beta (0.25 mg/ml) contains 20mM HEPES (pH6.0), 0.5M NaCl, 10% glycerol. |