missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL16 / SR-PSOX Protein
A DNA sequence encoding the Mouse CXCL16 / SR-PSOX, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10261-ec-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_075647.3 | |
CXCL16 / SR-PSOX Protein | |
Research Use Only | |
Cxcl16 | |
The biological activity determined by a chemotaxis bioassay using murine lymphocytes is in a concentration of 20-1000 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
66102 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLP | |
Greater than 98% as determined by SDS-PAGE and HPLC. |