missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CDNF Protein
A cDNA sequence encoding the CDNF was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10548-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
CDNF Protein | |
Research Use Only | |
CDNF Mouse is able to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons when immobilized at 5-30 microg/mL on a nitrocellulose-coated microplate. | |
Mouse | |
Untagged | |
CDNF protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
QGLEAGVGPR ADCEVCKEFL DRFYNSLLSR GIDFSADTIE KELLNFCSDA KGKENRLCYY LGATTDAATK ILGEVTRPMS VHIPAVKICE KLKKMDSQIC ELKYGKKLDL ASVDLWKMRV AELKQILQRW GEECRACAEK SDYVNLIREL APKYVEIYPQ TEL | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |