missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL4 / MIP1B Protein
A DNA sequence encoding the Mouse CCL4 / MIP1B, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10264-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_038680.1 | |
CCL4 / MIP1B Protein | |
Research Use Only | |
CCL4 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 20-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20303 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APMGSDPPTS CCFSYTSRQL HRSFVMDYYE TSSLCSKPAV VFLTKRGRQI CANPSEPWVT EYMSDLELN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |