missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque Amyloid protein 1
A DNA sequence encoding the Rhesus Macaque Amyloid protein A, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10247-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Amyloid protein 1 | |
Research Use Only | |
SAA1 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 10-100 ng/ml. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
RSWFSFLGEA YDGARDMWRA YSDMKEANYK NSDKYFHARG NYDAAQRGPG GVWAAEVISD ARENIQKLLG RGAEDTLADQ AANEWGRSGK DPNHFRPAGL PEKY | |
Greater than 97% as determined by SDS-PAGE and HPLC. |